Ethyl 5,6-dimethoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

49406-500MG 49406-1G 49406-5G
77335-536EA 357.41 USD
77335-536 77351-470 77375-756
Ethyl 5,6-dimethoxynicotinate ≥97%
Ethyl 5,6-dimethoxynicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29913989
CAS Number: 2387136-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR