3-Chloro-5-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

49388-500MG 49388-1G 49388-25G 49388-5G
77327-952EA 287.37 USD
77327-952 77351-466 77386-620 77375-754
3-Chloro-5-iodo-2-methoxypyridine ≥97%
3-Chloro-5-iodo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClINO
Storage Temperature: Ambient
MDL Number: MFCD18415715
CAS Number: 1261685-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR