3-Chloro-2-methoxy-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

49387-500MG 49387-1G 49387-5G 49387-25G
77327-950EA 168.92 USD
77327-950 77346-264 77364-270 77386-618
3-Chloro-2-methoxy-5-(methylthio)pyridine ≥97%
3-Chloro-2-methoxy-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNOS
MW: 189.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530008
CAS Number: 2734774-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR