(3-Bromo-2-chloro-5-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

49368-500MG 49368-1G 49368-5G
77335-462EA 354.32 USD
77335-462 77351-460 77375-748
(3-Bromo-2-chloro-5-methoxyphenyl)boronic acid ≥97%
(3-Bromo-2-chloro-5-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrClO₃
MW: 265.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756578
CAS Number: 2408112-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR