(3-Ethyl-4-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

49346-500MG 49346-1G 49346-5G 49346-25G
77327-938EA 142.14 USD
77327-938 77344-970 77364-260 77386-610
(3-Ethyl-4-fluorophenyl)methanol ≥97%
(3-Ethyl-4-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD17676554
CAS Number: 1135253-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR