N'-((5-Bromo-4-methylpyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

49336-500MG 49336-1G 49336-5G 49336-25G
77327-934EA 234.84 USD
77327-934 77351-452 77364-252 77386-608
N'-((5-Bromo-4-methylpyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%
N'-((5-Bromo-4-methylpyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrN₃O₂S
MW: 368.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902819
CAS Number: 2586233-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR