1-Bromo-3-fluoro-5-(4-fluorobenzyl)benzene ≥97%

Supplier: AOBChem USA

Warning

49334-500MG 49334-1G 49334-25G 49334-5G
77327-932EA 298.7 USD
77327-932 77351-450 77386-606 77375-736
1-Bromo-3-fluoro-5-(4-fluorobenzyl)benzene ≥97%
1-Bromo-3-fluoro-5-(4-fluorobenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂
Storage Temperature: Ambient
MDL Number: MFCD32902818
CAS Number: 2586126-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR