1-Bromo-3-fluoro-5-(4-methoxybenzyl)benzene ≥95%

Supplier: AOBChem USA

Warning

49333-500MG 49333-1G 49333-5G
77336-300EA 385.22 USD
77336-300 77351-448 77375-734
1-Bromo-3-fluoro-5-(4-methoxybenzyl)benzene ≥95%
1-Bromo-3-fluoro-5-(4-methoxybenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD32902817
CAS Number: 2586127-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR