1-Benzyl-3-bromo-5-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

49331-500MG 49331-1G 49331-5G
77327-930EA 318.27 USD
77327-930 77351-444 77375-730
1-Benzyl-3-bromo-5-fluorobenzene ≥95%
1-Benzyl-3-bromo-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD32902815
CAS Number: 875783-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR