4-(3,5-Dibromobenzyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49330-500MG 49330-1G 49330-5G
77336-154EA 379.04 USD
77336-154 77351-442 77375-728
4-(3,5-Dibromobenzyl)benzaldehyde ≥97%
4-(3,5-Dibromobenzyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀Br₂O
MW: 354.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902814
CAS Number: 2586125-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR