1,3-Dibromo-5-(4-methoxybenzyl)benzene ≥97%

Supplier: AOBChem USA

Warning

49328-500MG 49328-1G 49328-5G
77327-926EA 318.27 USD
77327-926 77351-438 77375-726
1,3-Dibromo-5-(4-methoxybenzyl)benzene ≥97%
1,3-Dibromo-5-(4-methoxybenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂Br₂O
MW: 356.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902812
CAS Number: 2586126-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR