1,3-Dibromo-5-(4-methylbenzyl)benzene ≥95%

Supplier: AOBChem USA

Warning

49327-500MG 49327-1G 49327-5G
77336-390EA 387.28 USD
77336-390 77351-436 77375-724
1,3-Dibromo-5-(4-methylbenzyl)benzene ≥95%
1,3-Dibromo-5-(4-methylbenzyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂Br₂
Storage Temperature: Ambient
MDL Number: MFCD32902811
CAS Number: 2586126-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR