1-Benzyl-3,5-dibromobenzene ≥97%

Supplier: AOBChem USA

Warning

49326-500MG 49326-1G 49326-5G
77335-506EA 356.38 USD
77335-506 77351-434 77375-722
1-Benzyl-3,5-dibromobenzene ≥97%
1-Benzyl-3,5-dibromobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀Br₂
Storage Temperature: Ambient
MDL Number: MFCD20269975
CAS Number: 71572-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR