2,6-Dichloro-3-(4-fluorobenzyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

49324-500MG 49324-1G 49324-5G 49324-25G
77327-924EA 280.16 USD
77327-924 77351-432 77364-248 77386-602
2,6-Dichloro-3-(4-fluorobenzyl)pyridine ≥95%
2,6-Dichloro-3-(4-fluorobenzyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂FN
MW: 256.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902810
CAS Number: 58597-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR