2,6-Dichloro-3-(4-methoxybenzyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

49323-500MG 49323-1G 49323-5G 49323-25G
77327-922EA 280.16 USD
77327-922 77351-430 77364-246 77386-600
2,6-Dichloro-3-(4-methoxybenzyl)pyridine ≥97%
2,6-Dichloro-3-(4-methoxybenzyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₁Cl₂NO
MW: 268.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902809
CAS Number: 58597-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR