3-Bromo-5-(4-methoxybenzyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

49318-500MG 49318-1G 49318-5G 49318-25G
77327-916EA 223.51 USD
77327-916 77351-424 77364-240 77386-594
3-Bromo-5-(4-methoxybenzyl)pyridine ≥97%
3-Bromo-5-(4-methoxybenzyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrNO
MW: 278.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902805
CAS Number: 2586126-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR