4-Bromo-5-chloro-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49254-500MG 49254-1G 49254-5G 49254-25G
77327-914EA 85.49 USD
77327-914 77342-214 77364-234 77375-716
4-Bromo-5-chloro-2-fluorobenzaldehyde ≥97%
4-Bromo-5-chloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26142710
CAS Number: 1603584-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR