2-Chloro-5-fluoro-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

49252-500MG 49252-1G 49252-5G
77339-742EA 622.12 USD
77339-742 77351-422 77375-714
2-Chloro-5-fluoro-4-methylphenol ≥97%
2-Chloro-5-fluoro-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD22414677
CAS Number: 1803588-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR