3-Chloro-5-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49249-500MG 49249-1G 49249-5G
77336-152EA 379.04 USD
77336-152 77351-420 77375-712
3-Chloro-5-fluoro-4-methylbenzaldehyde ≥97%
3-Chloro-5-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD28618322
CAS Number: 1547047-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR