2-Fluoro-1-isopropoxy-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49221-500MG 49221-1G 49221-25G 49221-5G
77327-908EA 298.7 USD
77327-908 77351-418 77386-590 77375-708
2-Fluoro-1-isopropoxy-3-methoxybenzene ≥97%
2-Fluoro-1-isopropoxy-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
MW: 184.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092195
CAS Number: 1369918-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR