3-Fluoro-N,4-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49217-500MG 49217-1G 49217-5G 49217-25G
77327-906EA 89.61 USD
77327-906 77342-364 77364-228 77375-706
3-Fluoro-N,4-dimethylbenzamide ≥97%
3-Fluoro-N,4-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO
MW: 167.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11767632
CAS Number: 1090829-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR