N-Cyclopropyl-3-fluoro-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49216-500MG 49216-1G 49216-5G 49216-25G
77327-904EA 89.61 USD
77327-904 77342-362 77364-226 77375-704
N-Cyclopropyl-3-fluoro-4-methylbenzamide ≥97%
N-Cyclopropyl-3-fluoro-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO
MW: 193.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10660164
CAS Number: 1090936-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR