N-Cyclohexyl-3-fluoro-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49215-500MG 49215-1G 49215-5G 49215-25G
77327-902EA 89.61 USD
77327-902 77342-360 77364-224 77375-702
N-Cyclohexyl-3-fluoro-4-methylbenzamide ≥97%
N-Cyclohexyl-3-fluoro-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO
MW: 235.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08883033
CAS Number: 923120-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR