3-Fluoro-N-methoxy-N,4-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49213-500MG 49213-1G 49213-5G 49213-25G
77327-898EA 126.69 USD
77327-898 77344-116 77364-220 77375-698
3-Fluoro-N-methoxy-N,4-dimethylbenzamide ≥97%
3-Fluoro-N-methoxy-N,4-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18840251
CAS Number: 774239-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR