(3-Fluoro-4-methylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49212-500MG 49212-1G 49212-5G 49212-25G
77327-896EA 89.61 USD
77327-896 77342-356 77364-218 77375-696
(3-Fluoro-4-methylphenyl)(morpholino)methanone ≥97%
(3-Fluoro-4-methylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNO₂
MW: 223.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21367432
CAS Number: 1455333-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR