(3-Fluoro-4-methylphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49211-1G 49211-5G 49211-25G
77341-666EA 98.88 USD
77341-666 77364-216 77375-694
(3-Fluoro-4-methylphenyl)(thiomorpholino)methanone ≥97%
(3-Fluoro-4-methylphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNOS
MW: 239.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13532551
CAS Number: 1197790-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR