2,5-Dibromo-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

49207-500MG 49207-1G 49207-5G
77336-150EA 379.04 USD
77336-150 77351-414 77375-692
2,5-Dibromo-3-methoxyphenol ≥97%
2,5-Dibromo-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆Br₂O₂
MW: 281.929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33421287
CAS Number: 1782369-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR