5-Bromo-1-chloro-3-fluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

49206-1G 49206-5G 49206-25G
77340-688EA 60.77 USD
77340-688 77364-212 77375-690
5-Bromo-1-chloro-3-fluoro-2-methylbenzene ≥95%
5-Bromo-1-chloro-3-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
Storage Temperature: Ambient
MDL Number: MFCD21603712
CAS Number: 1806058-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR