(3-Bromo-5-ethyl-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49200-500MG 49200-1G 49200-5G 49200-25G
77327-892EA 184.37 USD
77327-892 77347-202 77364-208 77386-586
(3-Bromo-5-ethyl-2-fluorophenyl)(methyl)sulfane ≥97%
(3-Bromo-5-ethyl-2-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀BrFS
Storage Temperature: Ambient
MDL Number: MFCD34564135
CAS Number: 2708196-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR