5-Bromo-4-methyl-2-phenoxypyridine ≥97%

Supplier: AOBChem USA

Warning

49189-500MG 49189-1G 49189-5G 49189-25G
77327-888EA 109.18 USD
77327-888 77343-218 77364-204 77375-686
5-Bromo-4-methyl-2-phenoxypyridine ≥97%
5-Bromo-4-methyl-2-phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06254605
CAS Number: 610279-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR