5-Bromo-2-(cyclopropylmethoxy)-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

49185-500MG 49185-1G 49185-5G 49185-25G
77327-886EA 173.04 USD
77327-886 77346-430 77364-200 77386-582
5-Bromo-2-(cyclopropylmethoxy)-4-methylpyridine ≥97%
5-Bromo-2-(cyclopropylmethoxy)-4-methylpyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
MW: 242.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31613894
CAS Number: 1600268-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR