5-Bromo-2-(4-chlorophenoxy)-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

49184-500MG 49184-1G 49184-5G 49184-25G
77327-884EA 141.11 USD
77327-884 77344-812 77364-198 77386-580
5-Bromo-2-(4-chlorophenoxy)-3-methylpyridine ≥97%
5-Bromo-2-(4-chlorophenoxy)-3-methylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₉BrClNO
MW: 298.563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25366218
CAS Number: 1882244-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR