5-Bromo-2-(3-chlorophenoxy)-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

49183-500MG 49183-1G 49183-5G 49183-25G
77327-882EA 141.11 USD
77327-882 77344-810 77364-196 77386-578
5-Bromo-2-(3-chlorophenoxy)-3-methylpyridine ≥97%
5-Bromo-2-(3-chlorophenoxy)-3-methylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₉BrClNO
MW: 298.563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26337456
CAS Number: 1874815-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR