(4-(Benzyloxy)-2,6-difluorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49178-500MG 49178-1G 49178-5G
77336-682EA 391.4 USD
77336-682 77351-410 77375-682
(4-(Benzyloxy)-2,6-difluorophenyl)(thiomorpholino)methanone ≥97%
(4-(Benzyloxy)-2,6-difluorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₈H₁₇F₂NO₂S
MW: 349.395 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756559
CAS Number: 2764731-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR