(4-(Benzyloxy)-2,6-difluorophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49177-500MG 49177-1G 49177-5G
77335-396EA 353.29 USD
77335-396 77351-408 77375-680
(4-(Benzyloxy)-2,6-difluorophenyl)(morpholino)methanone ≥97%
(4-(Benzyloxy)-2,6-difluorophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₈H₁₇F₂NO₃
MW: 333.329 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756558
CAS Number: 2764728-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR