4-(Benzyloxy)-N-cyclopropyl-2,6-difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

49175-500MG 49175-1G 49175-5G
77336-680EA 391.4 USD
77336-680 77351-404 77375-676
4-(Benzyloxy)-N-cyclopropyl-2,6-difluorobenzamide ≥97%
4-(Benzyloxy)-N-cyclopropyl-2,6-difluorobenzamide

Precaución: for Research Use Only

Formula: C₁₇H₁₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34756556
CAS Number: 2764730-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR