(3-Bromo-2-fluoro-6-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

49165-500MG 49165-1G 49165-5G
77339-414EA 587.1 USD
77339-414 77351-396 77375-668
(3-Bromo-2-fluoro-6-methylphenyl)methanol ≥97%
(3-Bromo-2-fluoro-6-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD30731371
CAS Number: 1540795-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR