methyl(2,4,6-trifluorophenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49155-500MG 49155-1G 49155-25G 49155-5G
77327-878EA 294.58 USD
77327-878 77351-394 77386-576 77375-666
methyl(2,4,6-trifluorophenyl)sulfane ≥97%
methyl(2,4,6-trifluorophenyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅F₃S
Storage Temperature: Ambient
MDL Number: MFCD32902801
CAS Number: 104745-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR