Ethyl 4-ethoxy-2,3-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

49149-500MG 49149-1G 49149-5G
77336-728EA 393.46 USD
77336-728 77351-392 77375-664
Ethyl 4-ethoxy-2,3-dimethylbenzoate ≥95%
Ethyl 4-ethoxy-2,3-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31772934
CAS Number: 1368039-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR