4-Ethoxy-2,3-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

49145-500MG 49145-1G 49145-5G
77327-874EA 336.81 USD
77327-874 77351-390 77375-662
4-Ethoxy-2,3-dimethylphenol ≥97%
4-Ethoxy-2,3-dimethylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD14537960
CAS Number: 1216256-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR