3,4-Difluoro-2-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

49133-500MG 49133-1G 49133-5G 49133-25G
77327-870EA 234.84 USD
77327-870 77351-386 77364-190 77386-570
3,4-Difluoro-2-methoxybenzamide ≥97%
3,4-Difluoro-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MW: 187.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789258
CAS Number: 1807182-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR