3,4-Difluoro-2-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49132-500MG 49132-1G 49132-5G 49132-25G
77327-868EA 173.04 USD
77327-868 77346-448 77364-188 77386-568
3,4-Difluoro-2-methoxy-N-methylbenzamide ≥97%
3,4-Difluoro-2-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34564133
CAS Number: 2734772-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR