N-Cyclopropyl-3,4-difluoro-2-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

49131-500MG 49131-1G 49131-5G 49131-25G
77327-866EA 166.86 USD
77327-866 77346-200 77364-186 77386-566
N-Cyclopropyl-3,4-difluoro-2-methoxybenzamide ≥97%
N-Cyclopropyl-3,4-difluoro-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₂NO₂
MW: 227.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178565
CAS Number: 2643368-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR