3,4-Difluoro-N,2-dimethoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49129-500MG 49129-1G 49129-5G 49129-25G
77327-864EA 147.29 USD
77327-864 77345-202 77364-184 77386-564
3,4-Difluoro-N,2-dimethoxy-N-methylbenzamide ≥97%
3,4-Difluoro-N,2-dimethoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD34564132
CAS Number: 2734774-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR