(3,4-difluoro-2-methoxyphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49128-500MG 49128-1G 49128-5G 49128-25G
77327-862EA 166.86 USD
77327-862 77346-198 77364-182 77386-562
(3,4-difluoro-2-methoxyphenyl)(morpholino)methanone ≥97%
(3,4-difluoro-2-methoxyphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂NO₃
MW: 257.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178563
CAS Number: 2643367-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR