(3,4-difluoro-2-methoxyphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

49127-500MG 49127-1G 49127-5G 49127-25G
77327-860EA 135.96 USD
77327-860 77344-496 77364-180 77386-560
(3,4-difluoro-2-methoxyphenyl)(thiomorpholino)methanone ≥97%
(3,4-difluoro-2-methoxyphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂NO₂S
MW: 273.299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178562
CAS Number: 2643368-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR