(3,4-difluoro-2-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

49126-500MG 49126-1G 49126-5G 49126-25G
77327-858EA 147.29 USD
77327-858 77345-200 77364-178 77386-558
(3,4-difluoro-2-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(3,4-difluoro-2-methoxyphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆F₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34564131
CAS Number: 2734778-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR