N-Methoxy-N,2-dimethylbenzamide ≥95%

Supplier: AOBChem USA

Warning

49088-1G 49088-5G 49088-25G
77341-228EA 77.25 USD
77341-228 77364-170 77375-652
N-Methoxy-N,2-dimethylbenzamide ≥95%
N-Methoxy-N,2-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD02684309
CAS Number: 130250-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR