(4-Bromophenyl)(pyridin-3-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

49084-1G 49084-5G 49084-25G
77341-192EA 74.16 USD
77341-192 77364-166 77375-648
(4-Bromophenyl)(pyridin-3-yl)methanone ≥97%
(4-Bromophenyl)(pyridin-3-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD00044286
CAS Number: 14548-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR