6-Bromo-2-fluoro-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

49054-1G 49054-5G 49054-25G
77341-016EA 66.95 USD
77341-016 77364-164 77375-644
6-Bromo-2-fluoro-3-(methylthio)pyridine ≥97%
6-Bromo-2-fluoro-3-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNS
MW: 222.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529995
CAS Number: 2734775-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR